siRNA Details
| Gene Symbol: Nol8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271397.1 | NP_001258326.1 | 15, 16 | 3943 | ||
| GenBank mRNA | AK017551.1 | - | 8, 9 | 1017 | ||
| AK147172.1 | - | 15, 16 | 3679 | |||
| BC058699.1 | - | 15, 16 | 4099 | |||
| BC094941.1 | - | 7, 8 | 813 | |||
| BC125575.1 | - | 15, 16 | 3669 | 40130090 | ||
| BC125579.1 | - | 15, 16 | 3669 | 40130094 | ||

Nol8
Genomic Map