siRNA Details
| Gene Symbol: Nol8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271397.1 | NP_001258326.1 | 14 | 3810 | ||
| GenBank mRNA | AK017551.1 | - | 7 | 884 | ||
| AK147172.1 | - | 14 | 3546 | |||
| BC026402.1 | - | 3 | 427 | |||
| BC058699.1 | - | 14 | 3966 | |||
| BC094941.1 | - | 6 | 680 | |||
| BC125575.1 | - | 14 | 3536 | 40130090 | ||
| BC125579.1 | - | 14 | 3536 | 40130094 | ||

Nol8
Genomic Map