siRNA Details
| Gene Symbol: Kctd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082924.1 | - | 3 | 779 | ||
| AK159067.1 | - | 3 | 2192 | |||
| BC096370.1 | - | 3 | 619 | 3597940 | ||
| BC131971.1 | - | 2 | 589 | 40130248 | ||
| BC131975.1 | - | 2 | 589 | 40130252 | ||

Kctd6
Genomic Map