siRNA Details
| Gene Symbol: Alpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027808.1 | NP_082084.1 | 3 | 674 | ||
| GenBank mRNA | AK018401.1 | - | 3 | 644 | ||
| AK041315.1 | - | 3 | 661 | |||
| AK173191.1 | - | 3 | 674 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alpk1| Gene Symbol: Alpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027808.1 | NP_082084.1 | 3 | 674 | ||
| GenBank mRNA | AK018401.1 | - | 3 | 644 | ||
| AK041315.1 | - | 3 | 661 | |||
| AK173191.1 | - | 3 | 674 | |||