siRNA Details
| Gene Symbol: Arl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027843.1 | NP_082119.1 | 1 | 169 | ||
| GenBank mRNA | AK018621.1 | - | 1 | 170 | ||
| BC104368.1 | - | 1 | 130 | 40049874 | ||
| BC104369.1 | - | 1 | 130 | 40049875 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arl14| Gene Symbol: Arl14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027843.1 | NP_082119.1 | 1 | 169 | ||
| GenBank mRNA | AK018621.1 | - | 1 | 170 | ||
| BC104368.1 | - | 1 | 130 | 40049874 | ||
| BC104369.1 | - | 1 | 130 | 40049875 | ||