siRNA Details
| Gene Symbol: Tysnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272090.1 | NP_001259019.1 | 4 | 2180 | ||
| NM_001272091.1 | NP_001259020.1 | 3 | 2090 | |||
| NM_001272092.1 | NP_001259021.1 | 3 | 2035 | |||
| NM_027912.1 | NP_082188.1 | 4 | 2221 | |||
| GenBank mRNA | AK005069.1 | - | 4 | 2222 | ||
| AK154513.1 | - | 4 | 2204 | |||
| AK180715.1 | - | 1 | 191 | |||
| AK218578.1 | - | 1 | 191 | |||
| AV148467.1 | - | 1 | 220 | |||

Tysnd1
Genomic Map