siRNA Details
| Gene Symbol: Tmem50a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027935.2 | NP_082211.1 | 5, 6 | 400 | ||
| GenBank mRNA | AK014282.1 | - | 5, 6 | 470 | ||
| AK147069.1 | - | 5, 6 | 436 | |||
| AK152630.1 | - | 5, 6 | 484 | |||
| AK152654.1 | - | 5, 6 | 455 | |||
| AK157214.1 | - | 5, 6 | 473 | |||
| AK164989.1 | - | 5, 6 | 466 | |||
| AK165636.1 | - | 5, 6 | 449 | |||
| BC094256.1 | - | 5, 6 | 400 | 6820666 | ||

Tmem50a
Genomic Map