siRNA Details
| Gene Symbol: Fam178b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126046.1 | NP_001119518.1 | 12 | 1659 | ||
| NM_027957.1 | NP_082233.1 | 3 | 282 | |||
| GenBank mRNA | AK006300.1 | - | 3 | 283 | ||
| BC114576.1 | - | 3 | 201 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam178b| Gene Symbol: Fam178b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126046.1 | NP_001119518.1 | 12 | 1659 | ||
| NM_027957.1 | NP_082233.1 | 3 | 282 | |||
| GenBank mRNA | AK006300.1 | - | 3 | 283 | ||
| BC114576.1 | - | 3 | 201 | |||