siRNA Details
| Gene Symbol: 1700019O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027966.1 | NP_082242.1 | 1 | 1592 | ||
| GenBank mRNA | AK006136.1 | - | 1 | 1592 | ||
| BC100497.1 | - | 1 | 1579 | 30916756 | ||
| BC145753.1 | - | 2 | 1501 | 40131056 | ||

1700019O17Rik
Genomic Map