siRNA Details
| Gene Symbol: Fam219a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159583.1 | NP_001153055.1 | 6 | 680 | ||
| NM_027993.3 | NP_082269.2 | 6 | 647 | |||
| GenBank mRNA | AK009522.1 | - | 6 | 631 | ||
| AK134737.1 | - | 6 | 683 | |||
| BC147808.1 | - | 6 | 609 | 9087525 | ||
| BC147810.1 | - | 6 | 607 | 9087527 | ||
| BC147834.1 | - | 6 | 609 | 9087551 | ||
| BC147836.1 | - | 6 | 609 | 9087553 | ||
| BC172167.1 | - | 6 | 576 | 9054837 | ||
| BC172173.1 | - | 6 | 606 | 9054843 | ||

Fam219a
Genomic Map