siRNA Details
| Gene Symbol: Tmub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132118.1 | - | 4 | 1008 | ||
| BC016187.1 | - | 3 | 949 | |||
| BC029841.1 | - | 3 | 1266 | 4209499 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmub2| Gene Symbol: Tmub2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132118.1 | - | 4 | 1008 | ||
| BC016187.1 | - | 3 | 949 | |||
| BC029841.1 | - | 3 | 1266 | 4209499 | ||