siRNA Details
| Gene Symbol: Tdrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173744.4 | NP_776105.1 | 3 | 553 | ||
| GenBank mRNA | AK011462.1 | - | 1 | 85 | ||
| AK077810.1 | - | 3 | 467 | |||
| AK166875.1 | - | 3 | 464 | |||
| AK166972.1 | - | 3 | 460 | |||
| BC099613.1 | - | 3 | 458 | 30031671 | ||
| BC132465.1 | - | 3 | 466 | 40130742 | ||
| BC132467.1 | - | 3 | 466 | 40130744 | ||
| BC145286.1 | - | 3 | 490 | 9053840 | ||

Tdrp
Genomic Map