siRNA Details
| Gene Symbol: Fbxl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178624.6 | NP_848739.1 | 2, 3 | 115 | ||
| GenBank mRNA | AK039010.1 | - | 2, 3 | 115 | ||
| AK044693.1 | - | 2, 3 | 112 | |||
| AK045742.1 | - | 2, 3 | 102 | |||
| AK089994.1 | - | 2, 3 | 91 | |||
| AK141177.1 | - | 2, 3 | 105 | |||
| AK163472.1 | - | 2, 3 | 111 | |||
| BC096582.1 | - | 2, 3 | 101 | 3963599 | ||
| BC145665.1 | - | 2, 3 | 95 | 40130968 | ||
| BC145998.1 | - | 2, 3 | 95 | 40131301 | ||

Fbxl2
Genomic Map