siRNA Details
| Gene Symbol: Dbndd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170975.2 | NP_001164446.2 | 3 | 1419 | ||
| NM_028146.4 | NP_082422.3 | 4 | 1560 | |||
| GenBank mRNA | AK013173.1 | - | 4 | 1458 | ||
| AK028187.1 | - | 4 | 1500 | |||
| BC046622.1 | - | 3 | 1354 | 6389902 | ||

Dbndd1
Genomic Map