siRNA Details
| Gene Symbol: Eml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162996.1 | NP_001156468.1 | 3 | 203 | ||
| NM_028153.1 | NP_082429.1 | 3 | 203 | |||
| GenBank mRNA | AK005562.1 | - | 3 | 169 | ||
| BC055476.1 | - | 3 | 203 | 4218133 | ||

Eml2
Genomic Map