siRNA Details
| Gene Symbol: Cdpf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164625.1 | NP_001158097.1 | 5 | 1009 | ||
| NM_001164626.1 | NP_001158098.1 | 4 | 803 | |||
| NM_197998.2 | NP_932115.2 | 5 | 950 | |||
| GenBank mRNA | AK008763.1 | - | 2 | 637 | ||
| BC025858.1 | - | 5 | 912 | 5007877 | ||

Cdpf1
Genomic Map