siRNA Details
| Gene Symbol: Zfp566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152814.2 | NP_690027.1 | 4 | 1151 | ||
| GenBank mRNA | AB010309.1 | - | 1 | 54 | ||
| AK012367.1 | - | 4 | 1128 | |||
| BC019790.1 | - | 4 | 1038 | 5151196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp566| Gene Symbol: Zfp566 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152814.2 | NP_690027.1 | 4 | 1151 | ||
| GenBank mRNA | AB010309.1 | - | 1 | 54 | ||
| AK012367.1 | - | 4 | 1128 | |||
| BC019790.1 | - | 4 | 1038 | 5151196 | ||