siRNA Details
| Gene Symbol: Zfp493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028402.2 | NP_082678.2 | 3 | 291 | ||
| GenBank mRNA | AK013687.1 | - | 231 | |||
| AK140336.1 | - | 3 | 183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp493| Gene Symbol: Zfp493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028402.2 | NP_082678.2 | 3 | 291 | ||
| GenBank mRNA | AK013687.1 | - | 231 | |||
| AK140336.1 | - | 3 | 183 | |||