siRNA Details
Gene Symbol: Zfp493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028402.2 | NP_082678.2 | 2 | 175 | ||
GenBank mRNA | AK013687.1 | - | 115 | |||
AK140336.1 | - | 2 | 67 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028402.2 | NP_082678.2 | 2 | 175 | ||
GenBank mRNA | AK013687.1 | - | 115 | |||
AK140336.1 | - | 2 | 67 |