siRNA Details
| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175200.4 | NP_780409.2 | 10 | 1160 | ||
| GenBank mRNA | AK076617.1 | - | 10 | 1162 | ||
| BC049729.1 | - | 6 | 377 | |||
| BC118929.2 | - | 10 | 1126 | 40090362 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Als2cr11| Gene Symbol: Als2cr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175200.4 | NP_780409.2 | 10 | 1160 | ||
| GenBank mRNA | AK076617.1 | - | 10 | 1162 | ||
| BC049729.1 | - | 6 | 377 | |||
| BC118929.2 | - | 10 | 1126 | 40090362 | ||