siRNA Details
Gene Symbol: 2210408F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008857.1 | - | 6 | 514 | ||
BC021953.1 | - | 8 | 870 | |||
BC099512.1 | - | 7 | 629 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 2210408F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008857.1 | - | 6 | 514 | ||
BC021953.1 | - | 8 | 870 | |||
BC099512.1 | - | 7 | 629 |