siRNA Details
| Gene Symbol: Dppa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018002.1 | NP_001018002.1 | 2 | 151 | ||
| NM_028610.2 | NP_082886.2 | 2 | 151 | |||
| GenBank mRNA | AK033197.1 | - | 2 | 152 | ||
| BC064754.1 | - | 2 | 90 | 30024281 | ||
| BY731814.1 | - | 2 | 117 | |||

Dppa4
Genomic Map