siRNA Details
| Gene Symbol: Otud4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081164.1 | NP_001074633.1 | 4, 5 | 574 | ||
| NM_001256033.1 | NP_001242962.1 | 4, 5 | 574 | |||
| GenBank mRNA | BC138373.1 | - | 4, 5 | 546 | 8861393 | |
| BC145259.1 | - | 4, 5 | 546 | 9053812 | ||

Otud4
Genomic Map