siRNA Details
| Gene Symbol: Cpne4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028719.1 | NP_082995.1 | 9 | 1165 | ||
| GenBank mRNA | AK043692.1 | - | 9 | 1178 | ||
| AK191212.1 | - | 2 | 181 | |||
| BC043087.1 | - | 9 | 1165 | 6417611 | ||
| BC063081.1 | - | 9 | 1165 | 6417611 | ||

Cpne4
Genomic Map