siRNA Details
| Gene Symbol: Cpne4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028719.1 | NP_082995.1 | 6, 7 | 914 | ||
| GenBank mRNA | AK014396.1 | - | 6, 7 | 916 | ||
| AK043692.1 | - | 6, 7 | 927 | |||
| AK202965.1 | - | 4, 5 | 280 | |||
| BC043087.1 | - | 6, 7 | 914 | 6417611 | ||
| BC063081.1 | - | 6, 7 | 914 | 6417611 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001109003, XM_008766557, AM747281.

Cpne4
Genomic Map