siRNA Details
| Gene Symbol: Kbtbd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028974.1 | NP_083250.1 | 1 | 670 | ||
| GenBank mRNA | AK048611.1 | - | 1 | 563 | ||
| BC139793.1 | - | 2 | 550 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kbtbd13| Gene Symbol: Kbtbd13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028974.1 | NP_083250.1 | 1 | 670 | ||
| GenBank mRNA | AK048611.1 | - | 1 | 563 | ||
| BC139793.1 | - | 2 | 550 | |||