siRNA Details
| Gene Symbol: Ndufs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029272.3 | NP_083548.1 | 8 | 627 | ||
| GenBank mRNA | AK003132.1 | - | 8 | 628 | ||
| BC013503.1 | - | 8 | 604 | 4223916 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufs7| Gene Symbol: Ndufs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029272.3 | NP_083548.1 | 8 | 627 | ||
| GenBank mRNA | AK003132.1 | - | 8 | 628 | ||
| BC013503.1 | - | 8 | 604 | 4223916 | ||