siRNA Details
Gene Symbol: Zfp820 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029281.2 | NP_083557.2 | 5 | 1393 | ||
GenBank mRNA | BC019219.1 | - | 5 | 1431 | 5066012 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp820 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029281.2 | NP_083557.2 | 5 | 1393 | ||
GenBank mRNA | BC019219.1 | - | 5 | 1431 | 5066012 |