siRNA Details
| Gene Symbol: 1700011M02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005877.1 | - | 3 | 282 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700011M02Rik| Gene Symbol: 1700011M02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005877.1 | - | 3 | 282 | ||