siRNA Details
| Gene Symbol: Higd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080846.1 | NP_543122.1 | 2, 3 | 602 | ||
| GenBank mRNA | AK009942.1 | - | 2, 3 | 443 | ||
| BC005568.1 | - | 2, 3 | 602 | 3967350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Higd1b| Gene Symbol: Higd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080846.1 | NP_543122.1 | 2, 3 | 602 | ||
| GenBank mRNA | AK009942.1 | - | 2, 3 | 443 | ||
| BC005568.1 | - | 2, 3 | 602 | 3967350 | ||