siRNA Details
| Gene Symbol: Rab11fip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080813.2 | NP_001074282.1 | 4 | 2207 | ||
| GenBank mRNA | BC034201.1 | - | 1 | 305 | ||
| BC091001.1 | - | 3 | 1646 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab11fip1| Gene Symbol: Rab11fip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080813.2 | NP_001074282.1 | 4 | 2207 | ||
| GenBank mRNA | BC034201.1 | - | 1 | 305 | ||
| BC091001.1 | - | 3 | 1646 | |||