siRNA Details
| Gene Symbol: Cep112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029586.2 | NP_083862.1 | 2 | 315 | ||
| NM_029606.3 | NP_083882.3 | 25 | 2862 | |||
| NM_145688.2 | NP_663721.2 | 4 | 537 | |||
| GenBank mRNA | AK005611.1 | - | 2 | 316 | ||
| AK006504.1 | - | 4 | 523 | |||
| BC005723.1 | - | 4 | 349 | |||
| BC048677.1 | - | 4 | 509 | |||
| BC086808.1 | - | 19 | 2102 | 30632964 | ||
| CA464637.1 | - | 4 | 539 | |||

Cep112
Genomic Map