siRNA Details
| Gene Symbol: Lsm8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133939.1 | NP_598700.1 | 4 | 333 | ||
| GenBank mRNA | AK008067.1 | - | 4 | 314 | ||
| AK088664.1 | - | 4 | 294 | |||
| AK154446.1 | - | 1198 | ||||
| AK167674.1 | - | 265 | ||||
| BC019458.1 | - | 4 | 333 | 4024697 | ||

Lsm8
Genomic Map