siRNA Details
Gene Symbol: Rbmxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029660.2 | NP_083936.1 | 1 | 1252 | ||
GenBank mRNA | AK005907.1 | - | 1 | 1253 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rbmxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029660.2 | NP_083936.1 | 1 | 1252 | ||
GenBank mRNA | AK005907.1 | - | 1 | 1253 |