siRNA Details
| Gene Symbol: Trdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029726.2 | NP_084002.2 | 3 | 512 | ||
| GenBank mRNA | AF223415.1 | - | 3 | 455 | ||
| AF223416.1 | - | 3 | 455 | |||
| AF223417.1 | - | 3 | 434 | |||
| AK009816.1 | - | 3 | 485 | |||
| BB658668.1 | - | 3 | 514 | |||
| BC034343.1 | - | 3 | 435 | 3589077 | ||

Trdn
Genomic Map