siRNA Details
| Gene Symbol: Spopl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165997.1 | NP_001159469.1 | 9 | 1207 | ||
| NM_001165998.1 | NP_001159470.1 | 8 | 1071 | |||
| NM_029773.2 | NP_084049.2 | 8 | 1110 | |||
| GenBank mRNA | AK014913.1 | - | 3 | 168 | ||
| AK162056.1 | - | 8 | 925 | |||
| BC111867.1 | - | 9 | 1081 | 40056321 | ||
| CD541905.1 | - | 3 | 204 | |||
| CF746115.1 | - | 5 | 564 | |||

Spopl
Genomic Map