siRNA Details
| Gene Symbol: 2610028H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029816.2 | NP_084092.2 | 7 | 999 | ||
| GenBank mRNA | AK011591.1 | - | 7 | 952 | ||
| AK161552.1 | - | 7 | 809 | |||
| BC051454.1 | - | 7 | 933 | 6308081 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610028H24Rik| Gene Symbol: 2610028H24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029816.2 | NP_084092.2 | 7 | 999 | ||
| GenBank mRNA | AK011591.1 | - | 7 | 952 | ||
| AK161552.1 | - | 7 | 809 | |||
| BC051454.1 | - | 7 | 933 | 6308081 | ||