siRNA Details
| Gene Symbol: Urah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029821.2 | NP_084097.1 | 3 | 489 | ||
| GenBank mRNA | AK004470.1 | - | 3 | 707 | ||
| AK013117.1 | - | 3 | 491 | |||
| BC051545.1 | - | 3 | 657 | 5255010 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Urah| Gene Symbol: Urah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029821.2 | NP_084097.1 | 3 | 489 | ||
| GenBank mRNA | AK004470.1 | - | 3 | 707 | ||
| AK013117.1 | - | 3 | 491 | |||
| BC051545.1 | - | 3 | 657 | 5255010 | ||