siRNA Details
| Gene Symbol: Ddrgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029832.2 | NP_084108.1 | 2 | 361 | ||
| GenBank mRNA | AK011170.1 | - | 2 | 292 | ||
| BC115616.2 | - | 2 | 361 | 40102754 | ||
| BC115617.1 | - | 2 | 361 | 40102757 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddrgk1| Gene Symbol: Ddrgk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029832.2 | NP_084108.1 | 2 | 361 | ||
| GenBank mRNA | AK011170.1 | - | 2 | 292 | ||
| BC115616.2 | - | 2 | 361 | 40102754 | ||
| BC115617.1 | - | 2 | 361 | 40102757 | ||