siRNA Details
| Gene Symbol: Slc9a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB089793.1 | - | 9, 10 | 876 | ||
| AF482993.1 | - | 8, 9 | 840 | |||
| AK049410.1 | - | 9, 10 | 872 | |||
| AK143281.1 | - | 9, 10 | 845 | |||
| AK160826.1 | - | 7, 8 | 714 | |||
| AK173069.1 | - | 9, 10 | 1212 | |||
| BC030879.1 | - | 9, 10 | 875 | 4241988 | ||
| BC034508.1 | - | 2, 3 | 276 | |||
| BC058947.1 | - | 9, 10 | 846 | 5702781 | ||

Slc9a8
Genomic Map