siRNA Details
| Gene Symbol: Tmem200a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029881.3 | NP_084157.2 | 2 | 1960 | ||
| GenBank mRNA | AK032569.1 | - | 2 | 1560 | ||
| AK048673.1 | - | 3 | 1355 | |||
| AK141454.1 | - | 1932 | ||||
| AK165025.1 | - | 2 | 1128 | |||
| AK173299.1 | - | 2 | 1960 | |||
| BC141266.1 | - | 2 | 1185 | 9055878 | ||
| BC141267.1 | - | 2 | 1185 | 9055879 | ||

Tmem200a
Genomic Map