siRNA Details
| Gene Symbol: Tmem50b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030018.3 | NP_084294.1 | 3 | 279 | ||
| GenBank mRNA | AK020977.1 | - | 3 | 284 | ||
| AK098154.1 | - | 3 | 243 | |||
| AK133966.1 | - | 3 | 307 | |||
| AK150773.1 | - | 267 | ||||
| AK151796.1 | - | 3 | 309 | |||
| AK152190.1 | - | 3 | 295 | |||
| AK152767.1 | - | 3 | 295 | |||
| AK155082.1 | - | 3 | 332 | |||
| AK170684.1 | - | 3 | 274 | |||
| AK172335.1 | - | 3 | 267 | |||
| BC004841.1 | - | 3 | 282 | 2649264 | ||

Tmem50b
Genomic Map