siRNA Details
| Gene Symbol: Tmem50b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030018.3 | NP_084294.1 | 3 | 258 | ||
| GenBank mRNA | AK020977.1 | - | 3 | 263 | ||
| AK098154.1 | - | 3 | 222 | |||
| AK133966.1 | - | 3 | 286 | |||
| AK150773.1 | - | 246 | ||||
| AK151796.1 | - | 3 | 288 | |||
| AK152190.1 | - | 3 | 274 | |||
| AK152767.1 | - | 3 | 274 | |||
| AK155082.1 | - | 3 | 311 | |||
| AK170684.1 | - | 3 | 253 | |||
| AK172335.1 | - | 3 | 246 | |||
| BC004841.1 | - | 3 | 261 | 2649264 | ||

Tmem50b
Genomic Map