siRNA Details
| Gene Symbol: Prr15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030024.2 | NP_084300.1 | 2 | 1330 | ||
| GenBank mRNA | AJ132433.2 | - | 2 | 1330 | ||
| AK136438.1 | - | 2 | 1510 | |||
| BC055838.1 | - | 2 | 1447 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr15| Gene Symbol: Prr15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030024.2 | NP_084300.1 | 2 | 1330 | ||
| GenBank mRNA | AJ132433.2 | - | 2 | 1330 | ||
| AK136438.1 | - | 2 | 1510 | |||
| BC055838.1 | - | 2 | 1447 | |||