siRNA Details
| Gene Symbol: Trappc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020026.1 | - | 5 | 592 | ||
| AK035837.1 | - | 1856 | ||||
| AK046610.1 | - | 5 | 620 | |||
| AK052819.1 | - | 1829 | ||||
| AK089083.1 | - | 5 | 433 | |||
| AK168294.1 | - | 6 | 880 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trappc6b| Gene Symbol: Trappc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020026.1 | - | 5 | 592 | ||
| AK035837.1 | - | 1856 | ||||
| AK046610.1 | - | 5 | 620 | |||
| AK052819.1 | - | 1829 | ||||
| AK089083.1 | - | 5 | 433 | |||
| AK168294.1 | - | 6 | 880 | |||