siRNA Details
| Gene Symbol: Asb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170710.1 | NP_001164181.1 | 4 | 1646 | ||
| NM_001170711.1 | NP_001164182.1 | 4 | 1652 | |||
| NM_030121.4 | NP_084397.3 | 4 | 1642 | |||
| GenBank mRNA | AK030168.1 | - | 4 | 1585 | ||
| BC025106.1 | - | 4 | 1566 | 4194829 | ||

Asb8
Genomic Map