siRNA Details
| Gene Symbol: Lsm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191004.1 | NP_001177933.1 | 4 | 342 | ||
| NM_030145.3 | NP_084421.1 | 3 | 243 | |||
| GenBank mRNA | AK005329.1 | - | 1 | 184 | ||
| AK019126.1 | - | 4 | 302 | |||
| AK078392.1 | - | 4 | 292 | |||
| AK141884.1 | - | 4 | 289 | |||
| AK158236.1 | - | 3 | 232 | |||
| AK161686.1 | - | 4 | 296 | |||
| BC030427.1 | - | 3 | 190 | 5370660 | ||
| BC098230.1 | - | 3 | 193 | 3372313 | ||
| BC099484.1 | - | 3 | 248 | 30936207 | ||
| DV648841.1 | - | 4 | 342 | |||

Lsm6
Genomic Map