siRNA Details
| Gene Symbol: 1500015O10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024283.3 | NP_077245.2 | 4 | 847 | ||
| GenBank mRNA | AK018770.1 | - | 4 | 602 | ||
| AK049805.1 | - | 4 | 848 | |||
| BC002254.1 | - | 4 | 780 | 3493553 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1500015O10Rik| Gene Symbol: 1500015O10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024283.3 | NP_077245.2 | 4 | 847 | ||
| GenBank mRNA | AK018770.1 | - | 4 | 602 | ||
| AK049805.1 | - | 4 | 848 | |||
| BC002254.1 | - | 4 | 780 | 3493553 | ||