siRNA Details
| Gene Symbol: Tnfrsf22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023680.3 | NP_076169.2 | 2, 3 | 201 | ||
| GenBank mRNA | AJ278265.1 | - | 2, 3 | 159 | ||
| AK012838.1 | - | 2, 3 | 202 | |||
| AY046551.1 | - | 2, 3 | 200 | |||
| AY165626.1 | - | 2, 3 | 224 | |||
| AY165627.1 | - | 2, 3 | 224 | |||
| BC076592.1 | - | 2, 3 | 215 | |||

Tnfrsf22
Genomic Map