siRNA Details
| Gene Symbol: Rilpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030259.1 | NP_084535.1 | 4 | 1168 | ||
| GenBank mRNA | AK133760.1 | - | 4 | 1300 | ||
| BC003324.1 | - | 4 | 1168 | 3156139 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rilpl2| Gene Symbol: Rilpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030259.1 | NP_084535.1 | 4 | 1168 | ||
| GenBank mRNA | AK133760.1 | - | 4 | 1300 | ||
| BC003324.1 | - | 4 | 1168 | 3156139 | ||